missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SGC 707

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SGC 707 is a potent allosteric inhibitor of protein arginine methyltransferase 3 (PRMT3) (IC50 = 31 nM). Inhibits PRMT3-mediated methyltransferase activity in vitro. Displays >100-fold selectivity against 31 other methyltransferases and 250 other non-epigenetic targets. Decreases H4R3me2a levels in HEK293 cells in vitro.
Negative control also available.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | SGC 707 |
| Purity | >98% |
| Molecular Weight (g/mol) | 298.34 |
| Inhibitors | SGC 707 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction