missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SGC 6870N

Product Code. 18024362
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18024362 25 mg 25mg
18024022 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18024362 Supplier Tocris Bioscience™ Supplier No. 7184/25
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Negative control for SGC 6870 (Cat. No. 7182)

SGC 6870N is a negative control for SGC 6870 (Cat. No. 7182). SGC 6870N is the (S)-enantiomer, which is inactive on PRMT6.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (S)-4-(5-Bromothiophene-2-carbonyl)-5-(3,5-dimethylphenyl)-7-methyl-1,3,4,5-tetrahydro-2H-benzo[e][1,4]diazepin-2-one
CAS 2561471-15-4
Quantity 25 mg
Target Protein Arginine Methyltransferase Inhibitors
Molecular Formula C23H21BrN2O2S
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.