missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ SGC 6870
Description
SGC 6870 is a potent and selective protein arginine methyltransferase 6 (PRMT6) allosteric inhibitor (IC50 = 77 nM). Exhibits selectivity over all other PRMTs and 23 methyltransferases. In HEK293T cells overexpressing PRMT6, inhibits asymmetric dimethylation of H3R2 (IC50 = 0.8 μM) Negative control SGC 6870N (Cat. No. 7184) also available.
Specifications
Specifications
| Chemical Name or Material | (R)-4-(5-Bromothiophene-2-carbonyl)-5-(3,5-dimethylphenyl)-7-methyl-1,3,4,5-tetrahydro-2H-benzo[e][1,4]diazepin-2-one |
| CAS | 2561471-27-8 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | Protein Arginine Methyltransferase Inhibitors |
| Molecular Formula | C23H21BrN2O2S |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?