missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SG2NA Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94911-0.1ml
This item is not returnable.
View return policy
Description
SG2NA Polyclonal antibody specifically detects SG2NA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| SG2NA | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin | |
| Cell cycle autoantigen SG2NA, cell cycle S/G2 nuclear autoantigen, GS2NA, S/G2 antigen, SG2NAnuclear autoantigen, striatin, calmodulin binding protein 3, striatin-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 200-460 of human STRN3 (NP_001077362.1). VRSLLGLSNSEPNGSVETKNLEQILNGGESPKQKGQEIKRSSGDVLETFNFLENADDSDEDEENDMIEGIPEGKDKHRMNKHKIGNEGLAADLTDDPDTEEALKEFDFLVTAEDGEGAGEARSSGDGTEWDKDDLSPTAEVWDVDQGLISKLKEQYKKERKGKKGVKRANRTKLYDMIADLGDDELPHIPSGIINQSRSASTRMTDHEGARAEEAEPITFPSGGGKSFIMGSDDVLLSVLGLGDLADLTVTNDADYSYDLP | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Neuroscience, Neurotransmission, Signal Transduction | |
| 29966 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction