missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFXN5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553303
This item is not returnable.
View return policy
Description
Immunogen sequence: ELEEGIDVLD SDGNLVGSSK IAARHALLE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Transports citrate; potential iron transporter. Classified as a mitochondrial tricarboxylate carrier protein that has citrate transport activity.
Specifications
| SFXN5 | |
| Polyclonal | |
| Unconjugated | |
| Sfxn5 | |
| BBG-TCC; C230001H08Rik; SFXN5; sideroflexin 5; sideroflexin-5; Tricarboxylate carrier BBG-TCC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 94097 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TD22 | |
| Sfxn5 | |
| Recombinant protein corresponding to Human SFXN5. Recombinant protein control fragment (Product #RP-91217). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction