missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFXN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15985-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| SFXN2 | |
| Polyclonal | |
| Unconjugated | |
| SFXN2 | |
| SFXN2, sideroflexin 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 118980, 294011, 94279 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q925N2, Q96NB2 | |
| Sfxn2 | |
| SFXN2 Fusion Protein Ag8788 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction