missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFXN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12296-1-AP-20UL
This item is not returnable.
View return policy
Description
Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).Specifications
| SFXN1 | |
| Polyclonal | |
| Unconjugated | |
| SFXN1 | |
| SFXN1, sideroflexin 1, TCC, Tricarboxylate carrier protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14057, 94081 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q99JR1, Q9H9B4 | |
| SFXN1 | |
| SFXN1 Fusion Protein Ag2985 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction