missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFXN1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68024-1-IG-150UL
This item is not returnable.
View return policy
Description
The SFXN1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SFXN1. This antibody recognizes human, rat, mouse antigen. The SFXN1 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| SFXN1 | |
| Monoclonal | |
| 1000 μg/mL | |
| FLJ12876 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Rat, Mouse | |
| 32 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 2D9F1 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 94081 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu