Learn More
Invitrogen™ SFTPD Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA579989
Product Code. 15935555
  • £400.00 / 100µg
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Lung Tissue, Rat Brain Tissue, PANC whole cell. IHC: human lung cancer tissue IHC-F: human placenta tissue. ICC/IF: A549 Cell, Hela Cell.
Surfactant protein D (SP-D) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular 'head' regions linked by triple-helical, collagen-like, strands. This group also includes SP-A and the serum proteins mannan-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SFTPD
Polyclonal
Unconjugated
SFTPD
AI573415; COLEC7; Collectin7; collectin-7; CP4; EGK_19692; Lung surfactant protein D; PSPD; PSP-D; PSP-D Surfactant protein D; pulmonary surfactant apoprotein; Pulmonary surfactant associated protein PSP-D; pulmonary surfactant protein D; pulmonary surfactant-associated protein D; SFTP 4; Sftp4; SFTPD; SPD; SP-D; surfactant associated protein D; Surfactant associated protein pulmonary 4; surfactant protein D; surfactant, pulmonary-associated protein D; surfactant-associated protein, pulmonary 4
Rabbit
Antigen affinity chromatography
RUO
25350, 6441
-20°C
Lyophilized
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
500 μg/mL
PBS with 5mg BSA and 0.05mg sodium azide
P35247, P35248
SFTPD
A synthetic peptide corresponding to a sequence at the C-terminus of human Surfactant protein D (292-321aa RSAAENAALQQLVVAKNEAAFLSMTD SKTE).
100 μg
Primary
Human, Rat