missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFTPD Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA5149258
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Surfactant protein D (SP-D) is synthesized and secreted by lung epithelial cells. It belongs to group III of the family of C-type lectins and members of this group has overall structure consisting of multiple globular 'head' regions linked by triple-helical, collagen-like, strands. This group also includes SP-A and the serum proteins mannan-binding protein, conglutinin and collectin-43, all of which have been shown to bind to the C1q receptor found on a wide variety of cells. Both SP-D and SP-A have been shown to enhance oxygen radical production by alveolar macrophages.
Specifications
| SFTPD | |
| Polyclonal | |
| Unconjugated | |
| SFTPD | |
| AI573415; COLEC7; Collectin7; collectin-7; CP4; EGK_19692; Lung surfactant protein D; PSPD; PSP-D; PSP-D Surfactant protein D; pulmonary surfactant apoprotein; Pulmonary surfactant associated protein PSP-D; pulmonary surfactant protein D; pulmonary surfactant-associated protein D; SFTP 4; Sftp4; SFTPD; SPD; SP-D; surfactant associated protein D; Surfactant associated protein pulmonary 4; surfactant protein D; surfactant, pulmonary-associated protein D; surfactant-associated protein, pulmonary 4 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 20390, 25350 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P35248, P50404 | |
| SFTPD | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse SP-D, Ala20-Phe374. | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction