missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTPD, Mouse anti-Human, Clone: 2C10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006441-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: AGMKTYSHRTMPSACTLVMCSSVESGLPGRDGRDGREGPRGEKGDPGLPGAAGQAGMPGQAGPVGPKGDNGSVGEPGPKGDTGPSGPPGPPGVPGPAGREGPLGKQGNIGPQGKPGPKGEAGPKGEVGAPGMQGSAGARGLAGPKGERGVPGERGVPGNTGAAGSAGAMGPQGSPGARGPPGLKGDKGIPGDKGAKGESGLPDVASLRQQVEALQGQVQHLQAAFSQYKKVELFPNGQSVGEKIFKTAGFVKPFTEAQLLCTQAGGQLASPRSAAENAALQQLVVAKNEAAFLSMTDSKTEGKFTYPTGESLVYSNWAPGKPNDDGGSEDCVEIFTNGKWNDRACGEKRLVVCEFSpecifications
| SFTPD | |
| Monoclonal | |
| Unconjugated | |
| surfactant protein D | |
| COLEC7/PSP-D/SFTP4/SP-D | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 2C10 | |
| In 1x PBS, pH 7.4 | |
| BC022318 | |
| SFTPD | |
| SFTPD (AAH22318.1, 21 a.a. ∼ 375 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6441 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction