missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFTPC Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119603
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
This gene encodes the pulmonary-associated surfactant protein C (SPC), an extremely hydrophobic surfactant protein essential for lung function and homeostasis after birth. Pulmonary surfactant is a surface-active lipoprotein complex composed of 90% lipids and 10% proteins which include plasma proteins and apolipoproteins SPA, SPB, SPC and SPD. The surfactant is secreted by the alveolar cells of the lung and maintains the stability of pulmonary tissue by reducing the surface tension of fluids that coat the lung. Multiple mutations in this gene have been identified, which cause pulmonary surfactant metabolism dysfunction type 2, also called pulmonary alveolar proteinosis due to surfactant protein C deficiency, and are associated with interstitial lung disease in older infants, children, and adults. Alternatively spliced transcript variants encoding different protein isoforms have been identified.
Specifications
| SFTPC | |
| Polyclonal | |
| Unconjugated | |
| Sftpc | |
| BRICD6; BRICHOS domain containing 6; LOW QUALITY PROTEIN: pulmonary surfactant-associated protein C; pro-SpC; PSP C; PSP-C; pulmonary surfactant apoprotein-2 SP-C; pulmonary surfactant associated protein C; pulmonary surfactant protein C; pulmonary surfactant-associated protein C; Pulmonary surfactant-associated proteolipid SPL(Val); SFTP 2; SFTP2; Sftp-2; Sftpc; SMDP2; SP C; SP5; SPC; SP-C; surfactant associated protein C; surfactant protein C; Surfactant pulmonary-associated protein C; Surfactant, pulmonary-associated protein C; surfactant-associated protein; type I SP-C; type I surfactant protein C; type II SP-C; type II surfactant protein C | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20389, 50683, 6440 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.04 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P11685, P11686, P21841 | |
| Sftpc | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human SFTPC. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction