missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTPB Precursor Monoclonal Antibody (SPB01 (1-2-A5-9)), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513975
This item is not returnable.
View return policy
Description
MA5-13975 targets Surfactant Protein B (Precursor) in IF and IHC (P) applications and shows reactivity with Human and Rat samples. The MA5-13975 immunogen is recombinant pro-surfactant protein B.
Pulmonary surfactant is a complex mixture of phospholipids and proteins that is secreted from type II cells in alveoli and reduces the surface tension at the alveolar air-liquid interface, providing alveolar stability necessary for normal ventillation. Four distinct proteins isolated from pulmonary surfactant are termed surfactant proteins A, B, C, and D. SP-A (28-36kDa) and SP-D (43kDa) are collagenous carbohydrate-binding proteins, whereas SP-B (8-9kDa) and SP-C (4kDa) are non-collagenous hydrophobic proteins. SP-B is expressed in pulmonary adenocarcinomas with acinar, papillary, bronchioloalveolar, and solid growth patterns. Squamous cell and large cell carcinomas of the lung and nonpulmonary adenocarcinomas do not express SP-B. ProSP-B is glycosylated in the Golgi apparatus and undergoes carboxy- and amino-terminal proteolysis by a cathepsin D-like protease.
Specifications
| SFTPB Precursor | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P07988, P22355 | |
| SFTPB | |
| Recombinant pro-surfactant protein B | |
| 500 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin) | |
| SPB01 (1-2-A5-9) | |
| Unconjugated | |
| SFTPB | |
| 18 kDa pulmonary-surfactant protein; 6 kDa protein; PSP-B; Pulmonary surfactant-associated protein B; pulmonary surfactant-associated proteolipid SPL(Phe); SFTB3; SFTP3; SFTPB; SMDP1; SP-B; surfactant associated protein B; surfactant protein B; surfactant pulmonary-associated protein B; surfactant, pulmonary-associated protein B | |
| Mouse | |
| Protein A | |
| RUO | |
| 192155, 6439 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction