missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFTPA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115987
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Surfactant Protein A.
This gene is one of several genes encoding pulmonary-surfactant associated proteins (SFTPA) located on chromosome 10. Mutations in this gene and a highly similar gene located nearby, which affect the highly conserved carbohydrate recognition domain, are associated with idiopathic pulmonary fibrosis. The current version of the assembly displays only a single centromeric SFTPA gene pair rather than the two gene pairs shown in the previous assembly which were thought to have resulted from a duplication.
Specifications
| SFTPA2 | |
| Polyclonal | |
| Unconjugated | |
| SFTPA2 | |
| 35 kDa pulmonary surfactant-associated protein; Alveolar proteinosis protein; COLEC5; collectin 5; Collectin-5; PSAP; PSPA; PSP-A; Pulmonary surfactant-associated protein A2; SFTP1; SFTPA; SFTPA2; SFTPA2B; SP-2A; SP-2A beta; SP-2A gamma; SP-A; SPA2; SP-A2; SP-A2 alpha; SP-A2 delta; SPAII; surfactant protein A2; surfactant, pulmonary-associated protein A2A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 729238 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8IWL1 | |
| SFTPA2 | |
| A synthesized peptide derived from human SFTPA2(Accession Q8IWL1), corresponding to amino acid residues G43-E93. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction