missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTPA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11850-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a lung surfactant protein that is a member of a subfamily of C-type lectins called collectins. The encoded protein binds specific carbohydrate moieties found on lipids and on the surface of microorganisms. This protein plays an essential role in surfactant homeostasis and in the defense against respiratory pathogens. Mutations in this gene are associated with idiopathic pulmonary fibrosis. Alternate splicing results in multiple transcript variants.Specifications
| SFTPA1 | |
| Polyclonal | |
| Unconjugated | |
| SFTPA1 | |
| Alveolar proteinosis protein, COLEC4, Collectin 4, PSAP, PSP A, PSPA, SFTP1, SFTPA, SFTPA1, SFTPA1B, SP A, SP A1, Surfactant Protein A, surfactant protein A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 653509 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.53 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8IWL2 | |
| SFTPA1 | |
| SFTPA1 Fusion Protein Ag2417 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction