Learn More
SFTPA1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11850-1-AP-20UL
 View more versions of this product
Product Code. 16879193
  • £135.00 / 20µL
Description
This gene encodes a lung surfactant protein that is a member of a subfamily of C-type lectins called collectins. The encoded protein binds specific carbohydrate moieties found on lipids and on the surface of microorganisms. This protein plays an essential role in surfactant homeostasis and in the defense against respiratory pathogens. Mutations in this gene are associated with idiopathic pulmonary fibrosis. Alternate splicing results in multiple transcript variants.
Specifications
Specifications
SFTPA1
Polyclonal
Unconjugated
SFTPA1
Alveolar proteinosis protein, COLEC4, Collectin 4, PSAP, PSP A, PSPA, SFTP1, SFTPA, SFTPA1, SFTPA1B, SP A, SP A1, Surfactant Protein A, surfactant protein A1
Rabbit
Antigen Affinity Chromatography
RUO
653509
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IWL2
SFTPA1
SFTPA1 Fusion Protein Ag2417
20 μL