missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFTA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121491
This item is not returnable.
View return policy
Description
SFTA2 (Surfactant Associated 2) is a Protein Coding gene and a putative surfactant protein.
Specifications
| SFTA2 | |
| Polyclonal | |
| Unconjugated | |
| SFTA2 | |
| Surfactant Associated 2; Surfactant-Associated Protein 2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 389376 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q6UW10 | |
| SFTA2 | |
| Thr20-Thr78 (Accession Q6UW10), with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction