missing translation for 'onlineSavingsMsg'
Learn More

SFRS4 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18426395 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426395 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426395 Supplier Novus Biologicals Supplier No. NBP204143AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SFRS4 Polyclonal antibody specifically detects SFRS4 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRS4
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias Pre-mRNA-splicing factor SRP75, serine/arginine-rich splicing factor 4, SFRS4, Splicing factor, arginine/serine-rich 4SRP001LB, SRP75SR splicing factor 4
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 250 and 300 of human Pre-mRNA-Splicing Factor SRP75 using the numbering given in entry NP_005617.2 (GeneID 6429).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6429
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.