missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS3 Antibody (SR1309), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | SFRS3 |
|---|---|
| Clone | SR1309 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18623434
|
Novus Biologicals
NBP3-22008-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682835
|
Novus Biologicals
NBP3-22008-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SFRS3 Monoclonal antibody specifically detects SFRS3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
| SFRS3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 6428 | |
| IgG | |
| Affinity purified |
| SR1309 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Pre-mRNA-splicing factor SRP20, serine/arginine-rich splicing factor 3, SFRS3splicing factor, arginine/serine-rich, 20-kD, Splicing factor, arginine/serine-rich 3SRP20, SRp20 | |
| A synthesized peptide derived from human SFRS3 (Uniprot #: P84103) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title