missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS2B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | SFRS2B |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
SFRS2B Polyclonal specifically detects SFRS2B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SFRS2B | |
| Unconjugated | |
| RUO | |
| Pre-mRNA-splicing factor SRP46, serine/arginine-rich splicing factor 8, Splicing factor SRp46, Splicing factor, arginine/serine-rich 2Bsplicing factor, arginine/serine-rich, 46kD, SR splicing factor 8, SRP46SFRS2B | |
| SRSF8 | |
| IgG |
| Polyclonal | |
| Rabbit | |
| Human | |
| 10929 | |
| Synthetic peptides corresponding to SFRS2B(splicing factor, arginine/serine-rich 2B) The peptide sequence was selected from the middle region of SFRS2B. Peptide sequence YRESRYGGSHYSSSGYSNSRYSRYHSSRSHSKSGSSTSSRSASTSKSSSA. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title