missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The antibody has no cross reaction to SFRS2B.
SFRS2 is necessary for the splicing of pre-mRNA. The protein is required for formation of the earliest ATP-dependent splicing complex and interacts with spliceosomal components bound to both the 5′- and 3′-splice sites during spliceosome assembly. It also is required for ATP-dependent interactions of both U1 and U2 snRNPs with pre-mRNA. And it interacts with other spliceosomal components, via the RS domains, to form a bridge between the 5′- and 3′-splice site binding components, U1 snRNP and U2AF. It binds to purine-rich RNA sequences, either 5′-AGSAGAGTA-3′ (S=C or G) or 5′-GTTCGAGTA-3′ and can bind to beta-globin mRNA and commit it to the splicing pathway.
Specifications
Specifications
| Antigen | SFRS2 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SRSF2 |
| Gene Accession No. | Q01130, Q62093, Q6PDU1 |
| Gene Alias | PR264, Protein PR264, SC 35, SC35, SFRS2, SFRS2A, Splicing component, 35 kDa, Splicing factor SC35, SRp30b, SRSF2 |
| Gene Symbols | Srsf2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?