missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS18 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-79851AF750
This item is not returnable.
View return policy
Description
SFRS18 Polyclonal antibody specifically detects SFRS18 in Human samples. It is validated for Western Blot
Specifications
| SFRS18 | |
| Polyclonal | |
| Western Blot | |
| bA98I9.2, C6orf111, chromosome 6 open reading frame 111, DKFZp564B0769, FLJ14752, FLJ14992, FLJ90147, HSPC306, MGC104269, RP11-98I9.2, Serine/arginine-rich-splicing regulatory protein 130, serine-arginine-rich-splicing regulatory protein 130, Splicing factor, arginine/serine-rich 130, splicing factor, arginine/serine-rich 18, SR rich protein, SR-related protein, SR-rich protein, SRRP130, SRrp130FLJ14853 | |
| The immunogen recognized by this antibody maps to a region between residue 200 to 250 of human splicing factor, arginine/serine-rich 18 (splicing factor, arginine/serine-rich 130) using the numbering given in entry NP_056306.1 (GeneID 25957). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 25957 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction