missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | SFRS14 |
|---|---|
| Dilution | Western Blot 1:200 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30233030
|
Novus Biologicals
NBP3-35539-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231711
|
Novus Biologicals
NBP3-35539-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SFRS14 Polyclonal antibody specifically detects SFRS14 in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| SFRS14 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Arginine/serine-rich-splicing factor 14, DKFZp686M2322, DKFZp779L2418, KIAA0365arginine/serine-rich 14 splicing factor, putative splicing factor, arginine/serine-rich 14, SFRS14, Splicing factor, arginine/serine-rich 14FLJ21855, SRFS14, SURP and G patch domain containing 2, SURP and G-patch domain-containing protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 500-700 of human SFRS14 (NP_055699.2).,, Sequence:, EAVGLQDIAPSPAAFPNFEDSTLFGREYIDHLKAWLVSSGCPLQVKKAEPEPMREEEKMIPPTKPEIQAKAPSSLSDAVPQRADHRVVGTIDQLVKRVIEGSLSPKERTLLKEDPAYWFLSDENSLEYKYYKLKLAEMQRMSENLRGADQKPTSADCAVRAMLYSRAVRNLKKKLLPWQRRGLLRAQGLRGWKARRATTGT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:200 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 10147 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title