missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS10, Mouse anti-Human, Clone: 7A1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006434-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: LGVFGLSLYTTERDLREVFSKYGPIADVSIVYDQQSRRSRGFAFVYFENVDDAKEAKERANGMELDGRRIRVDFSITKRPSpecifications
| SFRS10 | |
| Monoclonal | |
| Unconjugated | |
| splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila) | |
| DKFZp686F18120/Htra2-beta/SRFS10/TRA2-BETA/TRA2B/TRAN2B | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, KnockDown, Western Blot | |
| 7A1 | |
| In 1x PBS, pH 7.4 | |
| NM_004593 | |
| SFRS10 | |
| SFRS10 (NP_004584, 120 a.a. ∼ 199 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6434 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction