missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRS1 293T Cell Overexpression Lysate (Denatured), Abnova
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
This gene encodes a member of the arginine/serine-rich splicing factor protein family, and functions in both constitutive and alternative pre-mRNA splicing. The protein binds to pre-mRNA transcripts and components of the spliceosome, and can either activate or repress splicing depending on the location of the pre-mRNA binding site.
- Splicing factor, arginine/serine-rich 1
- Transfected cell line: 293T
- Plasmid: pCMV-SFRS1 full-length
- Host: Human
- Theoretical molecular weight: 27.39kDa
- Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)
Western Blotting
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | SFRS1 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC010264 |
| Molecular Weight (g/mol) | 27.39 |
| pH Range | 6.8 |
| Plasmid | pCMV-SFRS1 full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-SFRS1 antibody (H00006426-B01) by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction