missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRP4 Recombinant Rabbit Monoclonal Antibody (7Z6Q1)

SFRP4 Recombinant Rabbit Monoclonal Antibody (ARC0923)
Brand: Invitrogen MA535460
This item is not returnable.
View return policy
Description
SFRP4 Recombinant Monoclonal Antibody for Western Blot
Specifications
| SFRP4 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6FHJ7, Q9JLS4, Q9Z1N6 | |
| SFRP4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 201-300 of human SFRP4 (Q6FHJ7). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 7Z6Q1 | |
| Unconjugated | |
| SFRP4 | |
| Ddc4; DDC-4 protein; Frizzled protein, human endometrium; frizzled related protein; frizzled-related protein; frizzled-related protein 4; FRP; FRP-4; FRPHE; MGC26498; secreted frizzled related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related protein 4; secreted frizzled-related sequence protein 4; Sfrp4; sFRP-4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 20379, 6424, 89803 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction