missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFRP4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15328-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody got 41-51 kDa and 55 kDa bands in western blotting maybe due to differential posttranslational protein modification.
Secreted frizzled-related protein 4 (SFRP4) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. The expression of SFRP4 in ventricular myocardium correlates with apoptosis related gene expression.Specifications
| SFRP4 | |
| Polyclonal | |
| Unconjugated | |
| SFRP4 | |
| FRP 4, FRPHE, sFRP 4, SFRP4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6424 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6FHJ7 | |
| SFRP4 | |
| SFRP4 Fusion Protein Ag6909 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction