missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SFRP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15644885
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15644885 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15644885 Supplier Invitrogen™ Supplier No. PA550962

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FRP 2 Polyclonal Antibody detects endogenous levels of FRP 2 protein.

This gene encodes a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. Methylation of this gene is a potential marker for the presence of colorectal cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SFRP2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SFRP2
Gene Accession No. P97299, Q96HF1
Gene Alias AI851596; FKSG12; FRP 2; FRP2; FRP-2; MGC128911; MGC153618; MGC53344; Protein SDF5; SARP 1; SARP1; SARP-1; SDF 5; Sdf5; SDF-5; secreted apoptosis related protein 1; secreted apoptosis-related protein 1; secreted frizzled related protein 2; secreted frizzled-related protein 2; secreted frizzled-related sequence protein 2; secreted frizzled-related sequence protein 5; sFRP 2; SFRP2; sFRP-2; Sfrp2 protein; stromal cell derived factor 5; testicular tissue protein Li 170; UNQ361/PRO697
Gene Symbols SFRP2
Host Species Rabbit
Immunogen Synthesized peptide derived from the Internal region of human FRP-2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20319, 310552, 6423
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.