missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SFRP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119647
This item is not returnable.
View return policy
Description
Positive Control: A549, SH-SY-5Y, rat testis tissue, human colon cancer tissue, human breast tissue, human placenta tissue, mouse small intestine tissue. Subcellular Location: Secreted.
Secreted frizzled-related protein 1 (SFRP1) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP1 and SFRP5 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP1 is expressed in several human tissues, with the highest levels in heart.
Specifications
| SFRP1 | |
| Polyclonal | |
| Unconjugated | |
| SFRP1 | |
| 2210415K03Rik; AW011917; AW107218; AW742929; frizzled-related protein; FRP; FRP1; FRP-1; FrzA; SARP2; SARP-2; Secreted apoptosis-related protein 2; secreted frizzled related protein 1; secreted frizzled-related protein 1; secreted frizzled-related sequence protein 1; SFRP1; sFRP-1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20377, 6422, 84402 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8C4U3, Q8N474, Q9R168 | |
| SFRP1 | |
| Synthetic peptide within N-terminal human SFRP1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction