missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sFRP-5 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-121AF488
This item is not returnable.
View return policy
Description
sFRP-5 Polyclonal antibody specifically detects sFRP-5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| sFRP-5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Stem Cells | |
| 6425 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Alexa Fluor 488 | |
| frizzled-related protein 1b, FRP-1b, FRP1B, SARP3, SARP-3, secreted apoptosis related protein 3, secreted apoptosis-related protein 3, secreted frizzled-related protein 5, sFRP-5 | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction