missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sFRP-4 Rabbit anti-Human, Mouse, Rat, Clone: 7Z6Q1, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £425.00
Specifications
| Antigen | sFRP-4 |
|---|---|
| Clone | 7Z6Q1 |
| Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18388865
|
Novus Biologicals
NBP3-16352-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18358251
|
Novus Biologicals
NBP3-16352-100UL |
100 μg |
£425.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
sFRP-4 Monoclonal antibody specifically detects sFRP-4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| sFRP-4 | |
| Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Frizzled protein, human endometrium, FRP-4, frpHE, FRPHEMGC26498, secreted frizzled-related protein 4, secreted frizzled-related protein 4; secreted frizzled-related protein 4, sFRP-4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 201-300 of human sFRP-4 (Q6FHJ7). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 7Z6Q1 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 6424 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title