missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sFRP-4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98217-100ul
This item is not returnable.
View return policy
Description
sFRP-4 Polyclonal antibody specifically detects sFRP-4 in Mouse samples. It is validated for ELISA
Specifications
| sFRP-4 | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| Frizzled protein, human endometrium, FRP-4, frpHE, FRPHEMGC26498, secreted frizzled-related protein 4, secreted frizzled-related protein 4; secreted frizzled-related protein 4, sFRP-4 | |
| Produced in rabbits immunized with purified, recombinant Mouse sFRP-4 (Accession#: NP_057896.1; Met 1-Ser 351) | |
| 100 μg | |
| Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway | |
| 6424 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction