missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sFRP-4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£231.00 - £470.00
Specifications
| Antigen | sFRP-4 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18419130
|
Novus Biologicals
NBP1-81866-25ul |
25 μL |
£231.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18263187
|
Novus Biologicals
NBP1-81866 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
sFRP-4 Polyclonal specifically detects sFRP-4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| sFRP-4 | |
| Polyclonal | |
| Rabbit | |
| Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 6424 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RHMPWNITRMPNHLHHSTQENAILAIEQYEELVDVNCSAVLRFFLCAMYAPICTLEFLHDPIKPCKSVCQRARDDCEPLMKMYNHSWPESLACDELPVYDRGVCISPEAIVTDLPEDVKWIDITPDMMVQERPLDVDCKR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Frizzled protein, human endometrium, FRP-4, frpHE, FRPHEMGC26498, secreted frizzled-related protein 4, secreted frizzled-related protein 4; secreted frizzled-related protein 4, sFRP-4 | |
| SFRP4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title