missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sFRP-4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20330
This item is not returnable.
View return policy
Description
sFRP-4 Polyclonal specifically detects sFRP-4 in Human samples. It is validated for Western Blot.
Specifications
| sFRP-4 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| Frizzled protein, human endometrium, FRP-4, frpHE, FRPHEMGC26498, secreted frizzled-related protein 4, secreted frizzled-related protein 4; secreted frizzled-related protein 4, sFRP-4 | |
| Rabbit | |
| 40 kDa | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers, Wnt Signaling Pathway | |
| 6424 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q6FHJ7 | |
| SFRP4 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human sFRP-4. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction