missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sFRP-2 Antibody (OTI6C2), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74151AF532
This item is not returnable.
View return policy
Description
sFRP-2 Monoclonal specifically detects sFRP-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| sFRP-2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SFRP2 | |
| Recombinant protein fragment corresponding to amino acids 25-295 of human sFRP-2 (NP_003004) produced in E.coli. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Immunohistochemistry | |
| OTI6C2 | |
| Immunohistochemistry | |
| FRP2, FRP-2secreted apoptosis related protein 1, SARP-1, SARP1sFRP-2, SDF-5, Secreted apoptosis-related protein 1, secreted frizzled-related protein 2 | |
| Mouse | |
| 0.1 mL | |
| 6423 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction