missing translation for 'onlineSavingsMsg'
Learn More

sFRP-1 Rabbit anti-Human, Mouse, Rat, Clone: 5I6V1, Novus Biologicals™

Product Code. 18368546
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368546 100 μg 100µL
18323543 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18368546 Supplier Novus Biologicals Supplier No. NBP316851100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

sFRP-1 Monoclonal antibody specifically detects sFRP-1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen sFRP-1
Applications Western Blot
Classification Monoclonal
Clone 5I6V1
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FRP1, FRP-1FrzA, FRPSARP-2, SARP2sFRP-1, Secreted apoptosis-related protein 2, secreted frizzled-related protein 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human sFRP-1 (Q8N474).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 6422
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.