missing translation for 'onlineSavingsMsg'
Learn More
Learn More
sFRP-1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-499DL594
This item is not returnable.
View return policy
Description
sFRP-1 Polyclonal antibody specifically detects sFRP-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| sFRP-1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| FRP1, FRP-1FrzA, FRPSARP-2, SARP2sFRP-1, Secreted apoptosis-related protein 2, secreted frizzled-related protein 1 | |
| sFRP-1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to a 12 aa region of human Sfrp1 protein. (Uniprot: Q8N474) | |
| 0.1 mL | |
| Apoptosis | |
| 6422 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction