missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFMBT2 Antibody (PCRP-SFMBT2-1B7), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24306MFV500
This item is not returnable.
View return policy
Description
SFMBT2 Monoclonal antibody specifically detects SFMBT2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| SFMBT2 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| KIAA1617, Scm-like with four mbt domains 2, scm-like with four MBT domains protein 2 | |
| Recombinant full-length human SFMBT2 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| PCRP-SFMBT2-1B7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 57713 | |
| Store at 4°C in the dark. | |
| IgG2c |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction