Learn More
Invitrogen™ SFMBT1 Recombinant Human Monoclonal Antibody (RAB-C396)

Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551842
Description
This antibody recognizes SFMBT1 (Scm-like with four MBT domains protein 1). It binds to a folded domain, amino acids 1-455. SFMBT1 is a histone-binding protein, which is part of various corepressor complexes. It mediates the recruitment of corepressor complexes to target genes, followed by chromatin compaction and repression of transcription and plays a role during myogenesis.
This gene shares high similarity with the Drosophila Scm (sex comb on midleg) gene. It encodes a protein which contains four malignant brain tumor repeat (mbt) domains and may be involved in antigen recognition.
Specifications
| SFMBT1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q9UHJ3 | |
| SFMBT1 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing SFMBT1 protein under non-denaturing conditions. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| F(ab) κ |
| ChIP Assay, ELISA, Mass Spectrometry | |
| RAB-C396 | |
| Unconjugated | |
| SFMBT1 | |
| 4930442N21Rik; 9330180L21Rik; AA536974; hSFMBT; Renal ubiquitous protein 1; RU1; Scm like with four mbt domains 1; Scm-like with four mbt domains 1; scm-like with four MBT domains protein 1; Scm-related gene containing four mbt domains; Scm-related gene containing four mbt domains 1; Scm-related gene product containing four mbt domains; SFMBT; SFMBT1; Smr; wu:fc12f05 | |
| Human | |
| Metal-chelate chromatography | |
| RUO | |
| 51460 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.