missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SFM4Transfx-293 Media
Promote transfection using lipofection and support growth of HEK 293 cultures with HyClone™ SFM4Transfx-293 Media, a serum-free, animal-derived component free media.
Brand: Cytiva SH30860.01
Code : 83
Additional Details : Weight : 0.52500kg
Description
- ADCF
- Supports high transfection efficiency
- Metabolically designed for high cell yield and recombinant protein/vector production
- Allows for direct or sequential adaptation
- Component traceability
- Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Specifications
cGMP | |
SFM4Transfx-293 without L-Glutamine (liquid), 500mL | |
No Glutamine, No Animal Derived Components |
2° to 8°C | |
Liquid | |
500 mL |