missing translation for 'onlineSavingsMsg'
Learn More

Cytiva SFM4Transfx-293 Media

Promote transfection using lipofection and support growth of HEK 293 cultures with HyClone™ SFM4Transfx-293 Media, a serum-free, animal-derived component free media.

Brand:  Cytiva SH30860.LS

Code : 83

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10257653

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • ADCF
  • Supports high transfection efficiency
  • Metabolically designed for high cell yield and recombinant protein/vector production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Specifications

Specifications

cGMP
SFM4Transfx-293 without L-Glutamine (liquid), 6 x 1000mL
No Glutamine, No Animal Derived Components
2° to 8°C
Liquid
6 x 1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers