missing translation for 'onlineSavingsMsg'
Learn More

SFI1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17100574 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17100574 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17100574 Supplier Thermo Scientific Supplier No. PA5112709

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SFI1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen SFI1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Sfi1
Gene Accession No. A8K8P3
Gene Alias homolog of yeast Sfi1; hSFI1; hSfi1p; KIAA0542; PISD; PPP1R139; protein phosphatase 1, regulatory subunit 139; protein SFI1 homolog; Sfi1; SFI1 centrin binding protein; Sfi1 homolog, spindle assembly associated; Sfi1 homolog, spindle assembly associated (yeast)
Gene Symbols Sfi1
Host Species Rabbit
Immunogen Recombinant Human Protein SFI1 homolog protein (1003-1242AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9814
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.