missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF3B5 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18419DL594
This item is not returnable.
View return policy
Description
SF3B5 Polyclonal antibody specifically detects SF3B5 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SF3B5 | |
| Polyclonal | |
| Western Blot | |
| MGC3133, Pre-mRNA-splicing factor SF3b 10 kDa subunit, SF3b10, SF3b5, splicing factor 3B subunit 5, splicing factor 3b, subunit 5, 10kDa, Ysf3 | |
| Recognizes region between residues Between Between 1 and 50.. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 83443 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction