missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF3B4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20326
This item is not returnable.
View return policy
Description
SF3B4 Polyclonal specifically detects SF3B4 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| SF3B4 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| Hsh49, pre-mRNA splicing factor SF3b 49 kDa subunit, Pre-mRNA-splicing factor SF3b 49 kDa subunit, SAP 49, SAP49MGC10828, SF3B49, SF3b50, spliceosomal protein, spliceosome-associated protein (U2 snRNP), Spliceosome-associated protein 49, splicing factor 3B subunit 4, splicing factor 3b, subunit 4, 49kD, splicing factor 3b, subunit 4, 49kDa | |
| Rabbit | |
| 44 kDa | |
| 0.1 mL | |
| Neuroscience | |
| 10262 | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| 0.92mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q15427 | |
| SF3B4 | |
| Recombinant protein encompassing a sequence within the center region of human SF3B4. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Species reactivity based on sequence identity: Rat (100%), Bovine (100%), Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction