missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SF3B14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557077
This item is not returnable.
View return policy
Description
Immunogen sequence: YEDIFDAKNA CDHLSGFNVC NRYLVVLYYN ANRAFQKMDT KKKEEQLKLL KEKYGINTDP P Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 70%.
SF3B14 encodes a 14 kDa protein subunit of the splicing factor 3b complex. Splicing factor 3b associates with both the U2 and U11/U12 small nuclear ribonucleoprotein complexes (U2 snRNP) of spliceosomes. This 14 kDa protein interacts directly with subunit 1 of the splicing factor 3b complex. This 14 kDa protein also interacts directly with the adenosine that carries out the first transesterification step of splicing at the pre-mRNA branch site.
Specifications
| SF3B14 | |
| Polyclonal | |
| Unconjugated | |
| SF3B6 | |
| 0610009D07Rik; 6030419K15Rik; AV001342; CGI-110; HSPC175; Ht006; P14; pre mRNA branch site protein p14; pre-mRNA branch site protein p14; SAP14; SAP14a; SF3b 14 kDa subunit; Sf3b14; SF3B14a; SF3B6; spliceosome-associated protein, 14 kDa subunit; spliceosome-associated protein, 14-kDa; Splicing factor 3B subunit 6; splicing factor 3B, 14 kDa subunit; splicing factor 3B, 14 kDa subunit-like; splicing factor 3B, subunit 6; splicing factor 3b, subunit 6, 14kDa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51639 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry, Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y3B4 | |
| SF3B6 | |
| Recombinant protein corresponding to Human SF3B6. Recombinant protein control fragment (Product #RP-96001). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion