missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SF3A3 Recombinant Rabbit Monoclonal Antibody (23GB4775), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552741
This item is not returnable.
View return policy
Description
This gene encodes subunit 3 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 3 interacts with subunit 1 through its amino-terminus while the zinc finger domain of subunit 3 plays a role in its binding to the 15S U2 snRNP. This gene has a pseudogene on chromosome 20.
Specifications
| SF3A3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SF3A3 | |
| 4930512K19Rik; 60kDa; hypothetical protein LOC523250; LRRGT00093; pre-mRNA splicing factor SF3a (60kD); PRP9; PRPF9; SAP 61; Sap61; SF3A3; SF3a60; spliceosome associated protein 61; spliceosome-associated protein 61; splicesome-associated factor 61; splicing factor 3A subunit 3; splicing factor 3a subunit 3-like protein; splicing factor 3a, subunit 3; splicing factor 3a, subunit 3, 60kD; splicing factor 3a, subunit 3, 60kDa; wu:fi15a06; wu:fj94f01; zgc:110227 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10946, 313583, 75062 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4775 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q12874, Q9D554 | |
| SF3A3 | |
| A synthesized peptide derived from human SF3A3 (1-28AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction