missing translation for 'onlineSavingsMsg'
Learn More

SF3A2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18392154 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392154 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392154 Supplier Novus Biologicals Supplier No. NBP309943100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SF3A2 Polyclonal specifically detects SF3A2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SF3A2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS buffer, 2% sucrose
Gene Alias pre-mRNA splicing factor SF3A, subunit 2, PRP11, PRPF11, SAP 62, SAP62Prp11, SF3a66splicing factor 3a, subunit 2, 66kD, spliceosome associated protein 62, Spliceosome-associated protein 62, splicing factor 3A subunit 2, splicing factor 3a, subunit 2, 66kDa
Host Species Rabbit
Immunogen The immunogen is a synthetic peptide directed towards the N-terminal region of Human SF3A2. Peptide sequence GGKTGSGGVASSSESNRDRRERLRQLALETIDINKDPYFMKNHLGSYECK
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8175
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.