missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF3A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15106
This item is not returnable.
View return policy
Description
SF3A2 Polyclonal specifically detects SF3A2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| SF3A2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| pre-mRNA splicing factor SF3A, subunit 2, PRP11, PRPF11, SAP 62, SAP62Prp11, SF3a66splicing factor 3a, subunit 2, 66kD, spliceosome associated protein 62, Spliceosome-associated protein 62, splicing factor 3A subunit 2, splicing factor 3a, subunit 2, 66kDa | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8175 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.33mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Q15428 | |
| SF3A2 | |
| Recombinant protein encompassing a sequence within the center region of human SAP62. The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Human. Predict cross-reactivity based on sequence identity: Xenopus laevis (98%) and Xenopus tropicalis (98%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction