missing translation for 'onlineSavingsMsg'
Learn More

SF2523, MedChemExpress

Product Code. p-200097339
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494536 100 mg 100mg
16490729 1 mg 1mg
16476007 5 mg 5mg
16452706 25 mg 25mg
16449826 50 mg 50mg
16443288 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16494536 Supplier Medchem Express Supplier No. HY101146

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SF2523 is a highly selective and potent inhibitor of PI3K with IC50s of 34 nM, 158 nM, 9 nM, 241 nM and 280 nM for PI3Kα, PI3Kγ, DNA-PK, BRD4 and mTOR, respectively.

SF2523 is a highly selective and potent inhibitor of PI3K with IC50s of 34 nM, 158 nM, 9 nM, 241 nM and 280 nM for PI3Kα, PI3Kγ, DNA-PK, BRD4 and mTOR, respectively.

Specifications

Chemical Name or Material SF2523
CAS 1174428-47-7
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C19H17NO5S
Quantity 100 mg
Solubility Information DMSO : ≥ 30 mg/mL (80.77 mM)
SMILES O=C1C=C(N2CCOCC2)OC3=C1SC=C3C4=CC=C(OCCO5)C5=C4
Molecular Weight (g/mol) 371.41
Formula Weight 371.41
Percent Purity 97.32%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.