missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SF2523, MedChemExpress
Description
SF2523 is a highly selective and potent inhibitor of PI3K with IC50s of 34 nM, 158 nM, 9 nM, 241 nM and 280 nM for PI3Kα, PI3Kγ, DNA-PK, BRD4 and mTOR, respectively.
Specifications
Specifications
| Chemical Name or Material | SF2523 |
| CAS | 1174428-47-7 |
| Color | Earth Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C19H17NO5S |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 30 mg/mL (80.77 mM) |
| SMILES | O=C1C=C(N2CCOCC2)OC3=C1SC=C3C4=CC=C(OCCO5)C5=C4 |
| Molecular Weight (g/mol) | 371.41 |
| Formula Weight | 371.41 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?